Skip to Content
Merck
All Photos(1)

Key Documents

426091

Sigma-Aldrich

Methyl 2,5-dihydroxybenzoate

99%

Synonym(s):

Methyl gentisate

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
$53.50

$53.50


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 G
$53.50

About This Item

Linear Formula:
(HO)2C6H3CO2CH3
CAS Number:
Molecular Weight:
168.15
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$53.50


Check Cart for Availability

Request a Bulk Order

Assay

99%

mp

86-88 °C (lit.)

functional group

ester

SMILES string

COC(=O)c1cc(O)ccc1O

InChI

1S/C8H8O4/c1-12-8(11)6-4-5(9)2-3-7(6)10/h2-4,9-10H,1H3

InChI key

XGDPKUKRQHHZTH-UHFFFAOYSA-N

Related Categories

General description

Methyl 2,5-dihydroxybenzoate (methyl gentisate) is an alkyl ester of gentisic acid. It is reported to show less cytotoxic and mutagenic activity than hydroquinone with a potential to inhibit melanogenesis.[1] It has been synthesized from 2,5-dihydroxybenzoic acid. The crystal structure of the molecule was found to be planar.[2]

Application

Methyl 2,5-dihydroxybenzoate (methyl gentisate) may be used as a starting material in the synthesis of euonyminol.[3]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

E V Curto et al.
Biochemical pharmacology, 57(6), 663-672 (1999-02-26)
To discover safe and effective topical skin-lightening agents, we have evaluated alkyl esters of the natural product gentisic acid (GA), which is related to our lead compound methyl gentisate (MG), and four putative tyrosinase inhibitors, utilizing mammalian melanocyte cell cultures
Methyl 2, 5-dihydroxybenzoate.
Brown CL, et al.
Acta Crystallographica Section E, Structure Reports Online, 59(5), 630-631 (2003)
Total Synthesis of (.+-.)-Euonyminol, the Sesquiterpenoid Nucleus of Cathedulin K-19, via an Epoxide Cascade Cyclization.
White JD, et al.
Journal of the American Chemical Society, 117(38), 9780-9781 (1995)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service