Skip to Content
Merck
All Photos(1)

Documents

425648

Sigma-Aldrich

Trimethylgermanium iodide

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)3GeI
CAS Number:
Molecular Weight:
244.65
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:

Assay

98%

bp

134 °C (lit.)

density

1.815 g/mL at 25 °C (lit.)

SMILES string

C[Ge](C)(C)I

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Repr. 1B - Skin Corr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service