Skip to Content
Merck
All Photos(2)

Documents

425184

Sigma-Aldrich

Neodymium(III) trifluoromethanesulfonate

98%

Synonym(s):

Neodymium perfluoromethanesulfonate, Neodymium(III) triflate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Nd(CF3SO3)3
CAS Number:
Molecular Weight:
591.45
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:

Assay

98%

reaction suitability

core: neodymium
reagent type: catalyst

SMILES string

[Nd+3].[O-]S(=O)(=O)C(F)(F)F.[O-]S(=O)(=O)C(F)(F)F.[O-]S(=O)(=O)C(F)(F)F

InChI

1S/3CHF3O3S.Nd/c3*2-1(3,4)8(5,6)7;/h3*(H,5,6,7);/q;;;+3/p-3

InChI key

WYRSPTDNOIZOGA-UHFFFAOYSA-K

Application

Catalyst for:
  • Catalytic dehydration of fructose
  • Diels-Alder reactions of cyclopentadiene and alkyl acrylates
  • α-amination reactions of pyrazolones
  • Polymerization reactions of cyclosiloxanes
  • Direct polycondensation of dicarboxylic acids and diols
  • Regioselective reductive ring opening of benzylidene acetals
  • Paal-Knorr cyclocondensation
  • Nitration of benzene
A water-tolerant Lewis acid used in the Aldol reaction of silyl enol ethers with aldehydes.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The Journal of Organic Chemistry, 59, 3590-3590 (1994)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service