Skip to Content
Merck
All Photos(3)

Key Documents

424315

Sigma-Aldrich

3-(Chloromethyl)benzoic acid

97%

Synonym(s):

α-Chloro-m-toluic acid

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$40.10
25 G
$173.00

$40.10


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
$40.10
25 G
$173.00

About This Item

Linear Formula:
ClCH2C6H4CO2H
CAS Number:
Molecular Weight:
170.59
Beilstein:
637114
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$40.10


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

97%

mp

136-138 °C (lit.)

solubility

methanol: soluble 20 mg/mL, clear

functional group

carboxylic acid
chloro

SMILES string

OC(=O)c1cccc(CCl)c1

InChI

1S/C8H7ClO2/c9-5-6-2-1-3-7(4-6)8(10)11/h1-4H,5H2,(H,10,11)

InChI key

PBSUMBYSVFTMNG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

3-(Chloromethyl)benzoic acid (m-Chloromethylbenzoic acid) is a meta-substituted benzoic acid derivative. Correlations between Hammett substituent constants and calculated dipole moments and molecular transform and normalized molecular moment structure indices for m-chloromethylbenzoic acid was investigated. Hammett constants were observed to be linearly related only to the normalized charge moment index.[1]

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Molecular structural index control in property-directed clustering and correlation.
King JW and Molnar SP.
International Journal of Quantum Chemistry, 80(6), 1164-1171 (2000)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service