Skip to Content
Merck
All Photos(1)

Key Documents

424013

Sigma-Aldrich

1-(tert-Butoxycarbonyl)imidazole

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H12N2O2
CAS Number:
Molecular Weight:
168.19
EC Number:
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

bp

147-150 °C (lit.)

mp

45-47 °C (lit.)

SMILES string

CC(C)(C)OC(=O)n1ccnc1

InChI

1S/C8H12N2O2/c1-8(2,3)12-7(11)10-5-4-9-6-10/h4-6H,1-3H3

InChI key

MTBKGWHHOBJMHJ-UHFFFAOYSA-N

General description

1-(tert-butoxycarbonyl)imidazole is an imidazole derivative. Its gas-phase elimination kinetics study has been studied both theoretically and experimentally.

Application

1-(tert-butoxycarbonyl)imidazole was used in the preparation of water soluble polyrotaxane, via modification of α-cyclodextrin in the presence of KOH and N,N′-carbonyldiimidazole.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Block-Selective Movement of α-Cyclodextrins in Polyrotaxanes of PEI-b-PEG-b-PEI Copolymer.
Choi HS, et al.
Macromolecules, 38(23), 9878-9881 (2005)
Joint theoretical and experimental study of the gas-phase elimination kinetics of tert-butyl ester of carbamic, N,N-dimethylcarbamic, N-hydroxycarbamic acids and 1-(tert-butoxycarbonyl)-imidazole.
Mora JR, et al.
Journal of Physical Organic Chemistry, 20(12), 1021-1031 (2007)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service