Skip to Content
Merck
All Photos(2)

Documents

423297

Sigma-Aldrich

Boc-Gly-OtBu

99%

Synonym(s):

Boc-glycine tert-butyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)3COC(O)NHCH2CO2C(CH3)3
CAS Number:
Molecular Weight:
231.29
Beilstein:
5002580
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

reaction suitability

reaction type: Boc solid-phase peptide synthesis

mp

66-68 °C (lit.)

application(s)

peptide synthesis

SMILES string

CC(C)(C)OC(=O)CNC(=O)OC(C)(C)C

InChI

1S/C11H21NO4/c1-10(2,3)15-8(13)7-12-9(14)16-11(4,5)6/h7H2,1-6H3,(H,12,14)

InChI key

ZDKFMHKWZATBMR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service