Skip to Content
Merck
All Photos(1)

Key Documents

422851

Sigma-Aldrich

Ethylene-d4

99 atom % D

Synonym(s):

Tetradeuteroethylene

Sign Into View Organizational & Contract Pricing

Select a Size

1 L
$850.00

$850.00


Check Cart for Availability

Request more information

Select a Size

Change View
1 L
$850.00

About This Item

Linear Formula:
CD2=CD2
CAS Number:
Molecular Weight:
32.08
Beilstein:
1733307
EC Number:
MDL number:
UNSPSC Code:
12352003
PubChem Substance ID:
NACRES:
NA.12

$850.00


Check Cart for Availability

Request more information

vapor density

0.97 (vs air)

Quality Level

vapor pressure

35.04 atm ( 20 °C)

isotopic purity

99 atom % D

Assay

98.5% (CP)

autoignition temp.

842 °F

bp

−104 °C (lit.)

mp

−169 °C (lit.)

mass shift

M+4

SMILES string

[2H]\C([2H])=C(/[2H])[2H]

InChI

1S/C2H4/c1-2/h1-2H2/i1D2,2D2

InChI key

VGGSQFUCUMXWEO-LNLMKGTHSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Packaging

1 L quantity is packaged in a 450 mL carbon steel lecture bottle with brass CGA 110/180 valve. Nominal gas pressure at 21ºC is 19 psig for I L. This pressures is slightly above atmospheric pressure. Care must be taken when extracting this product from the cylinder.
All gas volumes are quoted at Standard Temperature and Pressure (STP) and not at Normal Temperature and Pressure (NTP). One STP Liter is 7.7% more gas than one NTP Liter.
This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Recommended products

Control valve Z146943 (brass, 1/4" hose outlet) or Z187224 (SS, 1/4" compression fitting outlet)
PTFE sealing washers GCA inlet size 180 Z146994
PTFE sealing washers CGA inlet size 110 Z146986

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Gas 1A - Press. Gas Liquefied gas - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

2A - Gases

WGK

nwg

Flash Point(F)

-148.0 °F - closed cup

Flash Point(C)

-100 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Krina Mehta et al.
The Journal of antimicrobial chemotherapy, 74(10), 2994-3002 (2019-07-06)
Reduced ethambutol serum concentrations are commonly observed among TB patients co-infected with HIV and may lead to treatment failure. To perform a population pharmacokinetic study of ethambutol in HIV/TB patients, and to evaluate an intensified ethambutol weight-based dosing strategy to
J F Huertas-Pérez et al.
Food additives & contaminants. Part A, Chemistry, analysis, control, exposure & risk assessment, 36(1), 96-108 (2018-12-14)
The production of certified reference materials requires the application of highly accurate methods for characterisation. A gas chromatography-isotope dilution mass spectrometry method, setting ambitious performance criteria, was developed for eight selected pesticides in soybeans. Pressurised liquid extraction was followed by

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service