Skip to Content
Merck
All Photos(1)

Key Documents

422541

Sigma-Aldrich

Ethyl 5-methylsalicylate

98%

Synonym(s):

Ethyl 2-hydroxy-5-methylbenzoate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3C6H3(OH)CO2C2H5
CAS Number:
Molecular Weight:
180.20
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

refractive index

n20/D 1.522 (lit.)

bp

251 °C (lit.)

density

1.103 g/mL at 25 °C (lit.)

functional group

ester

SMILES string

CCOC(=O)c1cc(C)ccc1O

InChI

1S/C10H12O3/c1-3-13-10(12)8-6-7(2)4-5-9(8)11/h4-6,11H,3H2,1-2H3

InChI key

ZGYXABNSOOACGL-UHFFFAOYSA-N

General description

Ethyl 5-methylsalicylate (ethyl 2-hydroxy-5-methylbenzoate) is a hydroxyl benzoate derivative. One of the methods reported for its synthesis is the esterification of p-cresotinic acid with absolute alcohol. It is formed as an intermediate during the synthesis of 2-propoxy-5-methylbenzoic acid. Ethyl 2-hydroxy-5-methylbenzoate has been isolated as a minor by product in the AlCl3 promoted reaction of 2-methyl furan with ethyl propiolate.

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

98.6 °F - closed cup

Flash Point(C)

37 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis of 2-Propoxy-5-Methylbenzoic Acid.
Brauer GM and Simon L.
Synthesis, 66(4), 313-315 (1962)
Influence of Lewis Acids on the Diels-Alder Reaction. IV. Reaction of 2-Methyl-and 2-Phenylfuran with Ethyl Propiolate.
McCulloch AW and McInnes AG.
Canadian Journal of Chemistry, 49(19), 3152-3157 (1971)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service