Skip to Content
Merck
All Photos(2)

Key Documents

422150

Sigma-Aldrich

Triphenylsilanethiol

98%

Synonym(s):

Mercaptotriphenylsilane

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$148.00

$148.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
$148.00

About This Item

Linear Formula:
(C6H5)3SiSH
CAS Number:
Molecular Weight:
292.47
Beilstein:
2941369
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

$148.00


Check Cart for Availability

Request a Bulk Order

Assay

98%

mp

101-104 °C (lit.)

SMILES string

S[Si](c1ccccc1)(c2ccccc2)c3ccccc3

InChI

1S/C18H16SSi/c19-20(16-10-4-1-5-11-16,17-12-6-2-7-13-17)18-14-8-3-9-15-18/h1-15,19H

InChI key

MQDRKELSDPESDZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Triphenylsilanethiol, also known as triphenylsilyl mercaptan, is an organosilicon compound that is commonly used as a reagent in organic synthesis. It can be used as a reagent for palladium-catalyzed cross-coupling reactions, such as the Stille coupling and the Sonogashira coupling reactions.

Application

Triphenylsilanethiol can be used as:      
  • A reactant to synthesis β-hydroxymercaptans or β-dihydroxysulfides by ring opening of epoxides.      
  • An additive in the selective C–H bond functionalization of widely available trialkylamine scaffolds.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tetrahedron Letters, 34, 3363-3363 (1993)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service