Skip to Content
Merck
All Photos(1)

Documents

421731

Sigma-Aldrich

(S)-(+)-2-(Dibenzylamino)-3-phenyl-1-propanol

99%

Synonym(s):

N,N-Dibenzyl-L-phenylalaninol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH2CH[N(CH2C6H5)2]CH2OH
CAS Number:
Molecular Weight:
331.45
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

form

solid

optical activity

[α]20/D +8.0°, c = 4 in methanol

mp

72-74 °C (lit.)

SMILES string

OC[C@H](Cc1ccccc1)N(Cc2ccccc2)Cc3ccccc3

InChI

1S/C23H25NO/c25-19-23(16-20-10-4-1-5-11-20)24(17-21-12-6-2-7-13-21)18-22-14-8-3-9-15-22/h1-15,23,25H,16-19H2/t23-/m0/s1

InChI key

ZXNVOFMPUPOZDF-QHCPKHFHSA-N

Application

Building block for the synthesis of HIV protease inhibitors.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

D Scholz et al.
Journal of medicinal chemistry, 37(19), 3079-3089 (1994-09-16)
A convenient procedure for the synthesis of 2-heterosubstituted statine derivatives as novel building blocks in HIV-protease inhibitors has been developed. The synthesis starts with protected L-phenylalaninols, which were converted to gamma-amino alpha, beta-unsaturated esters in a one-pot procedure. A highly
Pierre L. Beaulieu et al.
The Journal of organic chemistry, 61(11), 3635-3645 (1996-05-31)
Enantiomerically pure N,N-dibenzyl-alpha-amino aldehydes reacted with (chloromethyl)lithium, generated in situ from bromochloromethane and lithium metal, to give predominantly erythro aminoalkyl epoxides. Treatment of the crude epoxides with aqueous hydrochloric acid gave crystalline (2S,3S)-N,N-dibenzylamino chlorohydrin hydrochlorides in 32-56% overall yield and
Michael E. Pierce et al.
The Journal of organic chemistry, 61(2), 444-450 (1996-01-26)
DMP 323, a potent HIV-1 protease inhibitor, has been synthesized by an efficient stereoselective process, amenable to large scale preparations. The core C(2) symmetric diol was synthesized by a stereoselective pinacol coupling of CBZ protected D-phenylalanine. Judicious selection of protecting
Liu, C. et al.
Organic Process Research & Development, 1, 45-45 (1997)
P Y Lam et al.
Journal of medicinal chemistry, 39(18), 3514-3525 (1996-08-30)
High-resolution X-ray structures of the complexes of HIV-1 protease (HIV-1PR) with peptidomimetic inhibitors reveal the presence of a structural water molecule which is hydrogen bonded to both the mobile flaps of the enzyme and the two carbonyls flanking the transition-state

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service