Skip to Content
Merck
All Photos(2)

Key Documents

421707

Sigma-Aldrich

Boc-Phe-OMe

98%

Synonym(s):

N-(tert-Butoxycarbonyl)-L-phenylalanine methyl ester, Boc-L-phenylalanine methyl ester

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
$154.00

$154.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 G
$154.00

About This Item

Linear Formula:
C6H5CH2CH[NHCO2C(CH3)3]CO2CH3
CAS Number:
Molecular Weight:
279.33
Beilstein:
3061910
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.22

$154.00


Check Cart for Availability

Request a Bulk Order

Assay

98%

optical activity

[α]22/D −4.0°, c = 2 in methanol

reaction suitability

reaction type: Boc solid-phase peptide synthesis

mp

36-40 °C (lit.)

application(s)

peptide synthesis

SMILES string

COC(=O)[C@H](Cc1ccccc1)NC(=O)OC(C)(C)C

InChI

1S/C15H21NO4/c1-15(2,3)20-14(18)16-12(13(17)19-4)10-11-8-6-5-7-9-11/h5-9,12H,10H2,1-4H3,(H,16,18)/t12-/m0/s1

InChI key

SDSWSVBXRBXPRL-LBPRGKRZSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Natashya Falcone et al.
Chembiochem : a European journal of chemical biology, 20(6), 838-845 (2018-12-01)
Nicotinamide and pyridine-containing conjugates have attracted a lot of attention in research as they have found use in a wide range of applications including as redox flow batteries and calcium channel blockers, in biocatalysis, and in metabolism. The interesting redox

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service