Skip to Content
Merck
All Photos(1)

Documents

42135

Sigma-Aldrich

1,5-Dinitronaphthalene

≥97.0% (HPLC)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C10H6(NO2)2
CAS Number:
Molecular Weight:
218.17
Beilstein:
527184
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

≥97.0% (HPLC)

form

powder

mp

213-216 °C (lit.)
214-217 °C

SMILES string

[O-][N+](=O)c1cccc2c(cccc12)[N+]([O-])=O

InChI

1S/C10H6N2O4/c13-11(14)9-5-1-3-7-8(9)4-2-6-10(7)12(15)16/h1-6H

InChI key

ZUTCJXFCHHDFJS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

1,3-Dinitronaphthalene, 1,5-dinitronaphthalene and 1,8-dinitronaphthalene are the three dinitronaphthalene isomers. Their photocatalytic oxidation reactions in the presence of TiO2 Degussa P-25 grade has been investigated. 1,5-Dinitronaphthalene has been reported as environmental pollutant. Its genotoxic effects during in vivo wing somatic mutation and recombination test (SMART) of Drosophila melanogaster has been investigated. Delocalized intervalence radical anions of 1,5-dinitronaphthalene exhibits charge-transfer bands in their near-IR spectra. Crystals of 1,5-dinitronaphthalene are reported to be monoclinic with two molecules in each unit cell.

Application

1,5-Dinitronaphthalene has been employed as standard for the analysis of compounds of environmental interest by new nano-high-performance liquid chromatography-electron ionization mass spectrometry.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Eye Dam. 1 - Muta. 2 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kei Miyazawa et al.
Journal of mass spectrometry : JMS, 55(12), e4668-e4668 (2020-11-03)
Fragmentation of peptide radical cations [M]. + has been examined using matrix-assisted laser desorption/ionization (MALDI) in-source decay (ISD) with hydrogen-abstracting nitro-substituted matrices. The ISD spectra of peptides containing an arginine (Arg) residue at carboxyl (C)-termini showed preferential [w]+ ions when
Nano-high-performance liquid chromatography-electron ionization mass spectrometry approach for environmental analysis.
Cappiello A, et al.
Analytica Chimica Acta, 493(2), 125-136 (2003)
Yuki Hiruta et al.
The Analyst, 141(3), 910-917 (2015-12-10)
Temperature-responsive polymers incorporating molecular-recognition sites were developed as stationary phases for high-performance liquid chromatography (HPLC). The grafted stationary phases consisted of functional copolymers composed of N-isopropylacrylamide (NIPAAm) and N-acryloyl aromatic amino acid methyl esters, i.e., phenylalanine and tryptophan methyl esters
The crystal structure of 1, 5-dinitronaphthalene.
Trotter J.
Acta Crystallographica, 13(2), 95-99 (1960)
Stephen F Nelsen et al.
Journal of the American Chemical Society, 125(41), 12493-12501 (2003-10-09)
The Class III (delocalized) intervalence radical anions of 1,4-dinitrobenzene, 2,6-dinitronaphthalene, 2,6-dinitroanthracene, 9,9-dimethyl-2,7-dinitrofluorene, 4,4'-dinitrobiphenyl, and 1,5-dinitronaphthalene show charge-transfer bands in their near-IR spectra. The dinitroaromatic radical anions have comparable but slightly larger electronic interactions (H(ab) values) through the same aromatic bridges

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service