Skip to Content
Merck
All Photos(1)

Key Documents

421243

Sigma-Aldrich

Dimethyl cubane-1,4-dicarboxylate

99%

Synonym(s):

Cubane-1,4-dicarboxylic acid dimethyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H12O4
CAS Number:
Molecular Weight:
220.22
Beilstein:
1990376
MDL number:
UNSPSC Code:
12352103

Assay

99%

mp

163-165 °C (lit.)

InChI

1S/C12H12O4/c1-15-9(13)11-3-6-4(11)8-5(11)7(3)12(6,8)10(14)16-2/h3-8H,1-2H3

InChI key

OXBMFCGRQJNAOY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Tri-, tetra-, and pentacarbomethoxy cubane synthon.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Angewandte Chemie (International Edition in English), 32, 612-612 (1993)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service