Skip to Content
Merck
All Photos(1)

Key Documents

420719

Sigma-Aldrich

Lithium polysilicate solution

20 wt. % in H2O

Sign Into View Organizational & Contract Pricing

Select a Size

1 L
$68.80

$68.80


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 L
$68.80

About This Item

Linear Formula:
Li2Si5O11
CAS Number:
Molecular Weight:
330.30
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

$68.80


Check Cart for Availability

Request a Bulk Order

form

liquid

concentration

20 wt. % in H2O

density

1.16 g/mL at 25 °C

SMILES string

[Li+].[Li+].[O-][Si](=O)O[Si](=O)O[Si](=O)O[Si](=O)O[Si]([O-])=O

InChI key

WJZHQBRAMOCCLJ-UHFFFAOYSA-N

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service