Skip to Content
Merck
All Photos(1)

Key Documents

420131

Sigma-Aldrich

(+)-Diisopropyl-O,O′-bis(trimethylsilyl)-L-tartrate

99%, ChiraSelect reagent

Synonym(s):

(+)-O,O′-Bis(trimethylsilyl)-L-tartaric acid diisopropyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[-CH[OSi(CH3)3]CO2CH(CH3)2]2
CAS Number:
Molecular Weight:
378.61
Beilstein:
3620696
MDL number:
UNSPSC Code:
12352108
PubChem Substance ID:

product line

ChiraSelect

Assay

99%

optical activity

[α]20/D +57°, c = 1 in chloroform

refractive index

n20/D 1.428 (lit.)

bp

105-107 °C/1 mmHg (lit.)

density

0.972 g/mL at 25 °C (lit.)

SMILES string

CC(C)OC(=O)[C@H](O[Si](C)(C)C)[C@@H](O[Si](C)(C)C)C(=O)OC(C)C

InChI

1S/C16H34O6Si2/c1-11(2)19-15(17)13(21-23(5,6)7)14(22-24(8,9)10)16(18)20-12(3)4/h11-14H,1-10H3/t13-,14-/m1/s1

InChI key

WTAZEIUCUCCYIA-ZIAGYGMSSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

ChiraSelect is a trademark of Sigma-Aldrich Co. LLC

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

230.0 °F - closed cup

Flash Point(C)

110 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

David Holzberg et al.
The Journal of biological chemistry, 278(41), 40213-40223 (2003-07-02)
The transcription factor activator protein (AP)-1 plays crucial roles in proliferation, cell death, and the immune response. c-JUN is an important component of AP-1, but only very few c-JUN response genes have been identified to date. Activity of c-JUN is

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service