Skip to Content
Merck
All Photos(2)

Key Documents

419206

Sigma-Aldrich

Benzyl(triphenylphosphoranylidene)acetate

97%

Synonym(s):

(Benzyloxycarbonylmethylene)triphenylphosphorane, 2-(Triphenylphosphoranylidene)acetic acid phenylmethyl ester

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$84.50
25 G
$295.00

$84.50


0 left in stock

Request a Bulk Order

Select a Size

Change View
5 G
$84.50
25 G
$295.00

About This Item

Linear Formula:
(C6H5)3P=CHCO2CH2C6H5
CAS Number:
Molecular Weight:
410.44
MDL number:
UNSPSC Code:
12352108
PubChem Substance ID:
NACRES:
NA.22

$84.50


0 left in stock

Request a Bulk Order

Assay

97%

reaction suitability

reaction type: C-C Bond Formation

mp

120-122 °C (lit.)

functional group

ester
phenyl
phosphine

SMILES string

O=C(OCc1ccccc1)C=P(c2ccccc2)(c3ccccc3)c4ccccc4

InChI

1S/C27H23O2P/c28-27(29-21-23-13-5-1-6-14-23)22-30(24-15-7-2-8-16-24,25-17-9-3-10-18-25)26-19-11-4-12-20-26/h1-20,22H,21H2

InChI key

INKMLGJBBDRIQR-UHFFFAOYSA-N

Application

Reactant for:
  • Tributylphosphine-mediated vinylogous Wittig reactions
  • Synthesis of 1,2-dioxanes for antitrypanosomal activity
  • Organocatalytic Michael-type reactions / Wittig reactions of phosphorus ylides and unsaturated ketones
  • Stereoselective phosphine-catalyzed cycloaddition to form spirocyclopenteneoxindoles
  • Enantioselective synthesis of pantothenic acid
  • Preparation of phosphorus ylides from phosphoranes and acetic anhydride

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service