Skip to Content
Merck
All Photos(1)

Key Documents

41920

Sigma-Aldrich

2,5-Dinitrobenzoic acid

≥95.0% (HPLC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H4N2O6
CAS Number:
Molecular Weight:
212.12
Beilstein:
1983247
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Assay:
≥95.0% (HPLC)

Assay

≥95.0% (HPLC)

mp

178-182 °C

SMILES string

OC(=O)c1cc(ccc1[N+]([O-])=O)[N+]([O-])=O

InChI

1S/C7H4N2O6/c10-7(11)5-3-4(8(12)13)1-2-6(5)9(14)15/h1-3H,(H,10,11)

InChI key

YKMDNKRCCODWMG-UHFFFAOYSA-N

General description

2,5-Dinitrobenzoic acid is a benzoic acid derivative. Its proton, deuteron and proton spin-lattice relaxation times have been evaluated by using Schrödinger wave equation.[1]

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

L Latanowicz et al.
The journal of physical chemistry. A, 111(7), 1351-1357 (2007-02-01)
Temperature measurements of proton T1 (24.7 MHz), deuteron (deuterated hydroxyl group) T1 (55.2 MHz), and proton T1(rho) (B1 = 9 G) spin-lattice relaxation times of 2,5-dinitrobenzoic acid have been performed. An analysis of present experimental data together with previously published

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service