Skip to Content
Merck
All Photos(2)

Documents

416622

Sigma-Aldrich

Phenylsuccinic anhydride

99%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H8O3
CAS Number:
Molecular Weight:
176.17
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

Assay

99%

form

solid

bp

191-192 °C/12 mmHg (lit.)

mp

53-55 °C (lit.)

SMILES string

O=C1CC(C(=O)O1)c2ccccc2

InChI

1S/C10H8O3/c11-9-6-8(10(12)13-9)7-4-2-1-3-5-7/h1-5,8H,6H2

InChI key

HDFKMLFDDYWABF-UHFFFAOYSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ramzi Mukred Saeed et al.
Frontiers in bioengineering and biotechnology, 8, 4-4 (2020-02-11)
Chitosan nanoparticles (NPs) are widely studied as vehicles for drug, protein, and gene delivery. However, lack of sufficient stability, particularly under physiological conditions, render chitosan NPs of limited pharmaceutical utility. The aim of this study is to produce stable chitosan

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service