Skip to Content
Merck
All Photos(1)

Key Documents

415480

Sigma-Aldrich

Chloroquine diphosphate salt

97%

Synonym(s):

N4-(7-Chloro-4-quinolinyl)-N1,N1-dimethyl-1,4-pentanediamine diphosphate salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H26ClN3 · 2H3PO4
CAS Number:
Molecular Weight:
515.86
Beilstein:
4223142
EC Number:
MDL number:
UNSPSC Code:
12352100

Assay

97%

SMILES string

OP(O)(O)=O.OP(O)(O)=O.CCN(CC)CCCC(C)Nc1ccnc2cc(Cl)ccc12

InChI

1S/C18H26ClN3.2H3O4P/c1-4-22(5-2)12-6-7-14(3)21-17-10-11-20-18-13-15(19)8-9-16(17)18;2*1-5(2,3)4/h8-11,13-14H,4-7,12H2,1-3H3,(H,20,21);2*(H3,1,2,3,4)

InChI key

QKICWELGRMTQCR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

replaced by


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Margaret A L Blackie et al.
Bioorganic & medicinal chemistry letters, 20(3), 1078-1080 (2009-12-26)
Synthesis of the potent antiplasmodial 4-aminoquinoline, phenylequine (PQ), is reported for the first time. PQ and the two analogues show increased efficacy in moving from the chloroquine sensitive D10 to the chloroquine resistant K1 strain in vitro. The in vivo
Clive L Yeates et al.
Journal of medicinal chemistry, 51(9), 2845-2852 (2008-04-10)
A series of diaryl ether substituted 4-pyridones have been identified as having potent antimalarial activity superior to that of chloroquine against Plasmodium falciparum in vitro and murine Plasmodium yoelii in vivo. These were derived from the anticoccidial drug clopidol through
David J Bacon et al.
Antimicrobial agents and chemotherapy, 51(4), 1172-1178 (2007-01-16)
In vitro drug susceptibility testing with the malaria parasite has been used to assess the antimalarial activities of new compounds and to monitor drug resistance in field isolates. We investigated the validity of a SYBR green I fluorescent-based assay under
Onyeka Onyeibor et al.
Journal of medicinal chemistry, 48(7), 2701-2709 (2005-04-02)
A series of analogues of cryptolepine (1) have been synthesized and evaluated for their in vitro antiplasmodial and cytotoxic properties. The IC(50) values of several compounds (11a, 11k-m, 11o, 13) against Plasmodium falciparum (strain K1) were <0.1 muM, 5-10-fold lower
Gajanan Wanare et al.
Bioorganic & medicinal chemistry letters, 20(15), 4675-4678 (2010-06-26)
Both the lack of a credible malaria vaccine and the emergence and spread of parasites resistant to most of the clinically used antimalarial drugs and drug combination have aroused an imperative need to develop new drugs against malaria. In present

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service