Skip to Content
Merck
All Photos(1)

Documents

41308

Sigma-Aldrich

2,3,5-Tri-O-benzoyl-β-D-ribofuranosyl cyanide

purum, ≥98.0% (HPLC)

Synonym(s):

2,5-Anhydro-3,4,6-tri-O-benzoyl-D-allononitrile

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H21NO7
CAS Number:
Molecular Weight:
471.46
Beilstein:
1443499
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:
NACRES:
NA.22

grade

purum

Assay

≥98.0% (HPLC)

form

powder

SMILES string

O=C(OC[C@H]1O[C@@H](C#N)[C@H](OC(=O)c2ccccc2)[C@@H]1OC(=O)c3ccccc3)c4ccccc4

InChI

1S/C27H21NO7/c28-16-21-23(34-26(30)19-12-6-2-7-13-19)24(35-27(31)20-14-8-3-9-15-20)22(33-21)17-32-25(29)18-10-4-1-5-11-18/h1-15,21-24H,17H2/t21-,22+,23-,24+/m0/s1

InChI key

OFMYTVNDIRTGMR-UARRHKHWSA-N

Other Notes

Building block for C-nucleoside synthesis

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A. Dondoni et al.
Helvetica Chimica Acta, 85, 3331-3331 (2002)
M. Manferdini et al.
Tetrahedron, 58, 1005-1005 (2002)
P.J. Dudfield et al.
Journal of the Chemical Society. Perkin Transactions 1, 2937-2937 (1999)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service