Skip to Content
Merck
All Photos(1)

Key Documents

413062

Sigma-Aldrich

Phenyl trifluoroacetate

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CO2CF3
CAS Number:
Molecular Weight:
190.12
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

liquid

refractive index

n20/D 1.419 (lit.)

bp

146-147 °C (lit.)

density

1.276 g/mL at 25 °C (lit.)

functional group

ester
fluoro
phenoxy

SMILES string

FC(F)(F)C(=O)Oc1ccccc1

InChI

1S/C8H5F3O2/c9-8(10,11)7(12)13-6-4-2-1-3-5-6/h1-5H

InChI key

DVCMYAIUSOSIQP-UHFFFAOYSA-N

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

127.4 °F - closed cup

Flash Point(C)

53 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

pH Dependent effect of ?-cyclodextrin on the hydrolysis rate of trifluoroacetate esters.
Fernandez MA, et al.
The Journal of Organic Chemistry, 62(2), 7554-7559 (1997)
Kinetics and mechanisms of reactions of pyridines and imidazoles with phenyl acetates and trifluoroacetates in aqueous acetonitrile with low content of water: nucleophilic and general base catalysis in ester hydrolysis.
Neuvonen H.
J. Chem. Soc. Perkin Trans. II, 2(2), 159-167 (1987)
Synthesis, characterization, and copolymerization of a series of novel acid monomers based on sulfonimides for proton conducting membranes.
Rahman MK, et al.
Macromolecules, 37(15), 5572-5577 (2004)
1, 4-Addition of arylboronic acids and arylsiloxanes to a, ?-unsaturated carbonyl compounds via transmetalation to dicationic palladium (II) complexes.
Nishikata T, et al.
Organometallics, 23(18), 4317-4324 (2004)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service