Skip to Content
Merck
All Photos(3)

Documents

412546

Sigma-Aldrich

2-Amino-1-methylbenzimidazole

95%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H9N3
CAS Number:
Molecular Weight:
147.18
MDL number:
UNSPSC Code:
12352100
eCl@ss:
32151902
PubChem Substance ID:
NACRES:
NA.22

Assay

95%

SMILES string

Cn1c(N)nc2ccccc12

InChI

1S/C8H9N3/c1-11-7-5-3-2-4-6(7)10-8(11)9/h2-5H,1H3,(H2,9,10)

InChI key

XDFZKQJLNGNJAN-UHFFFAOYSA-N

General description

The carbonyl-scavenging ability of 2-amino-1-methylbenzimidazole has been investigated. Mechanism of Menshutkin reaction between 2-amino-1-methylbenzimidazole and iodomethane has been studied in gas phase and in liquid acetonitrile. It is reported to form adducts with natural allyl, phenethyl, and benzyl isothiocyanates.

Application

2-Amino-1-methylbenzimidazole may be used for the preparation of 2- or 3-carboxy-4H-pyrimido[2,1-b]-benzazol-4-ones and novel functionalized spiropyran′s derivatives of 2H-1,3-benzoxazinone series.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

André Melo et al.
The journal of physical chemistry. B, 110(4), 1877-1888 (2006-02-14)
The quaternization reaction between 2-amino-1-methylbenzimidazole and iodomethane was investigated in the gas phase and in liquid acetonitrile. Both experimental and theoretical techniques were used in this study. In the experimental part of this work, accurate second-order rate constants were obtained
Synthesis and structural characterization of novel 2-benzimidazolylthioureas: adducts of natural isothiocyanates and 2-amino-1-methylbenzimidazole.
Smiechowska A, et al.
Structural Chemistry, 21(5), 955-964 (2010)
Antony O Bulanov et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 71(3), 1146-1152 (2008-06-14)
Six novel functionalized spiropyran's derivatives of 2H-1,3-benzoxazinone series were synthesized by introducing the substituents with chelating ability into 2H-chromene part of the 8'-formyl-7'-hydroxy-3-methyl-4-oxo-3,4-dihydro-2H-1,3-benzoxazine-2-spiro-2'-[2H]-chromene (I) by condensation with 2-aminophenol, 2-amino-4-methylphenol, 2-amino-4-nitrophenol, 2-amino-1-methylbenzimidazole, 4-amino-4H-1,2,4-triazole, N-(4-aminophenyl)acetamide. (1)H NMR, UV/vis, IR spectroscopy combined with
J J Wade et al.
Journal of medicinal chemistry, 26(4), 608-611 (1983-04-01)
Reactions of 2-aminobenzothiazole, 2-aminobenzoxazole, and 2-amino-1-methylbenzimidazole with dimethyl aminofumarate (DMAF) or diethyl ethoxymethylenemalonate (DEEM) led to 2- or 3-carboxy-4H-pyrimido[2,1-b]-benzazol-4-ones, respectively. Subsequent derivatization of these carboxylic acids gave the corresponding tetrazolylcarboxamides and tetrazoles. These acidic compounds were tested in the rat
Paria Pashazadeh-Panahi et al.
Journal of molecular recognition : JMR, 33(5), e2830-e2830 (2020-02-19)
Ketoconazole or Nizoral is an antifungal medication used to treat various type of fungal infections. It has been reported that this antifungal agent is able to induce a variety of heart function side effects, such as long-QT syndrome, and ventricular

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service