Skip to Content
Merck
All Photos(1)

Key Documents

411132

Sigma-Aldrich

(R)-(−)-α-(Trifluoromethyl)benzyl alcohol

99%

Synonym(s):

(−)-Phenyl(trifluoromethyl)carbinol, (R)-(−)-1-Phenyl-2,2,2-trifluoroethanol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH(CF3)OH
CAS Number:
Molecular Weight:
176.14
Beilstein:
2413959
EC Number:
MDL number:
UNSPSC Code:
12352002
PubChem Substance ID:

Assay

99%

form

liquid

optical activity

[α]21/D −29°, c = 2.6 in chloroform

optical purity

ee: 97% (GLC)

refractive index

n20/D 1.459 (lit.)

bp

73-76 °C/9 mmHg (lit.)

mp

20 °C (lit.)

density

1.300 g/mL at 20 °C (lit.)

SMILES string

O[C@H](c1ccccc1)C(F)(F)F

InChI

1S/C8H7F3O/c9-8(10,11)7(12)6-4-2-1-3-5-6/h1-5,7,12H/t7-/m1/s1

InChI key

VNOMEAQPOMDWSR-SSDOTTSWSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Chiral solvating agent used for determination of enantiomeric composition by [1]H-NMR.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

188.6 °F - closed cup

Flash Point(C)

87 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Pirkle, W.H. and Rinaldi, P.L.
The Journal of Organic Chemistry, 42, 3217-3217 (1977)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service