Skip to Content
Merck
All Photos(2)

Key Documents

410233

Sigma-Aldrich

1H-Benzotriazole-1-methanol

98%

Synonym(s):

1-(Hydroxymethyl)benzotriazole, 1-Benzotriazolemethanol, NSC 12463

Sign Into View Organizational & Contract Pricing

Select a Size

10 G
$66.90
50 G
$232.00

$66.90


0 left in stock

Request a Bulk Order

Select a Size

Change View
10 G
$66.90
50 G
$232.00

About This Item

Empirical Formula (Hill Notation):
C7H7N3O
CAS Number:
Molecular Weight:
149.15
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

$66.90


0 left in stock

Request a Bulk Order

Quality Level

Assay

98%

reaction suitability

reaction type: C-C Bond Formation

mp

150-152 °C (lit.)

functional group

hydroxyl

SMILES string

OCn1nnc2ccccc12

InChI

1S/C7H7N3O/c11-5-10-7-4-2-1-3-6(7)8-9-10/h1-4,11H,5H2

InChI key

MXJIHEXYGRXHGP-UHFFFAOYSA-N

Application

Catalyst for:
  • Hydrolysis of phenyl esters of a-furoic acid

Used as:
  • Corrosion inhibitor of iron in aerated acidic media
  • Reactive oxygen scavenger

Reactant for:
  • Synthesis of hydroxy-skipped bis-homo-inositols as potential glycosidase inhibitors via Sharpless asymmetric dihydroxylation and substrate-directed anionic hydroxymethylation
  • Enantioselective synthesis of aplysin utilizing a palladium-catalyzed addition of an arylboronic acid to an allenic alcohol, followed by Eschenmoser/Claisen rearrangement
  • Synthesis of substituted dihydroquinoline carboxylic acids as antitumor and HIV-1 integrase inhibitors
  • Gosteli-Claisen rearrangement
Safe reagent for the in situ generation of anhydrous formaldehyde in organic solvents.[1]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gülsev Dilber et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 217, 128-140 (2019-04-01)
In this study, the novel 3-(1H-benzo[d][1,2,3]triazol-1-yl)methoxy substituted novel phthalonitrile compounds (2 and 6) were synthesized. Their non-peripheral and peripheral zinc (II) (3 and 7), manganese (III) (4 and 8) and copper (II) (5 and 9) phthalocyanine complexes were synthesized for
Organic Letters (2007)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service