Skip to Content
Merck
All Photos(1)

Key Documents

40902

Sigma-Aldrich

3-(Bromomethyl)phenyl isothiocyanate

≥95.0%

Synonym(s):

3-Isothiocyanatobenzyl bromide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H6BrNS
CAS Number:
Molecular Weight:
228.11
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

≥95.0%

reaction suitability

reagent type: cross-linking reagent

mp

35-38 °C

solubility

chloroform: 0.1 g/mL, clear, very faintly brownish-yellow

SMILES string

BrCc1cccc(c1)N=C=S

Application

Hetero-bifunctional linker specially for the quaternisation of pyridyl-nitrogen in order to introduce an amine or thiol reactive group. Synthetic building block for the preparation of fluorescent labels[1]

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Litak, P.T.;Kauffman, J.M.
Journal of Heterocyclic Chemistry, 31, 457-457 (1994)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service