Skip to Content
Merck
All Photos(1)

Key Documents

408581

Sigma-Aldrich

Z-Thr-OMe

98%

Synonym(s):

N-Carbobenzyloxy-L-threonine methyl ester, Z-L-Threonine methyl ester

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$151.00

$151.00


0 left in stock

Request a Bulk Order

Select a Size

Change View
5 G
$151.00

About This Item

Linear Formula:
CH3CH(OH)CH(NHCO2CH2C6H5)CO2CH3
CAS Number:
Molecular Weight:
267.28
Beilstein:
2220800
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.22

$151.00


0 left in stock

Request a Bulk Order

Assay

98%

optical activity

[α]25/D −18°, c = 1 in methanol

reaction suitability

reaction type: solution phase peptide synthesis

mp

92-94 °C (lit.)

application(s)

peptide synthesis

SMILES string

COC(=O)[C@@H](NC(=O)OCc1ccccc1)[C@@H](C)O

InChI

1S/C13H17NO5/c1-9(15)11(12(16)18-2)14-13(17)19-8-10-6-4-3-5-7-10/h3-7,9,11,15H,8H2,1-2H3,(H,14,17)/t9-,11+/m1/s1

InChI key

OPZWAOJFQFYYIX-KOLCDFICSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service