Skip to Content
Merck
All Photos(1)

Key Documents

407437

Sigma-Aldrich

1,6-Heptadiyne

97%

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$62.90

$62.90


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
$62.90

About This Item

Linear Formula:
HC≡C(CH2)3C≡CH
CAS Number:
Molecular Weight:
92.14
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$62.90


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

97%

form

liquid

refractive index

n20/D 1.441 (lit.)

bp

111.5 °C (lit.)

density

0.805 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

C#CCCCC#C

InChI

1S/C7H8/c1-3-5-7-6-4-2/h1-2H,5-7H2

InChI key

RSPZSDWVQWRAEF-UHFFFAOYSA-N

General description

Polymerization of 1,6-heptadiyne using an insoluble Zeigler-Natta catalyst affords soluble polymer having six-membered ring containing polyene.[1] Ruthenium(II)-catalyzed reaction of a substituted 1,6-heptadiyne with norbornene affords a tandem [2+ 2+ 2]/[4+2] cycloaddition product and a [2+ 2+2] cycloadduct.[2] Cyclocopolymerization of 1,6-heptadiyne with dipropargyl ether under nitrogen atmosphere using KSCN, KBr and KI as initiators in N,N-dimethyl formamide has been studied.[3]

Application

1,6-Heptadiyne may be used in the preparation of free-standing polymer films of poly(1,6-heptadiyne).[1]

Pictograms

Flame

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

48.2 °F - closed cup

Flash Point(C)

9 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Poly (1, 6-heptadiyne), a free-standing polymer film dopable to high electrical conductivity.
Gibson HW, et al.
Journal of the American Chemical Society, 105(13), 4417-4431 (1983)
Simultaneous synthesis and doping of poly (1, 6-heptadiyne-co-dipropargyl ether) using ionic initiators.
Sivakumar C, et al.
Polymer, 43(6), 1781-1787 (2002)
Ruthenium-catalyzed tandem [2+ 2+ 2]/[4+ 2] cycloaddition of 1, 6-heptadiyne with norbornene.
Yamamoto Y, et al.
Organometallics, 17(10), 1910-1912 (1998)
Alla V Lipeeva et al.
Molecules (Basel, Switzerland), 24(11) (2019-06-15)
Synthesis of 1,2,3-triazole-substituted coumarins and also 1,2,3-triazolyl or 1,2,3-triazolylalk-1-inyl-linked coumarin-2,3-furocoumarin hybrids was performed by employing the cross-coupling and copper catalyzed azide-alkyne cycloaddition reaction approaches. The synthesized compounds were evaluated for their in vitro antibacterial activity against Staphylococcusaureus, Bacilliussubtilis, Actinomycesviscosus and

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service