Skip to Content
Merck
All Photos(1)

Key Documents

406694

Sigma-Aldrich

Poly(propylene glycol) bis(2-aminopropyl ether)

average Mn ~4,000

Sign Into View Organizational & Contract Pricing

Select a Size

250 ML
$109.00

$109.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
250 ML
$109.00

About This Item

Linear Formula:
CH3CH(NH2)CH2[OCH2CH(CH3)]nNH2
CAS Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

$109.00


Check Cart for Availability

Request a Bulk Order

form

viscous liquid

mol wt

average Mn ~4,000

refractive index

n20/D 1.452

density

0.997 g/mL at 25 °C

SMILES string

CC(N)CO.CC(O)CO

InChI

1S/C4H9NO2/c6-3-1-5-2-4(3)7/h3-7H,1-2H2

InChI key

JCZPOYAMKJFOLA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 2

Flash Point(F)

440.1 °F - closed cup

Flash Point(C)

226.7 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service