Skip to Content
Merck
All Photos(1)

Key Documents

406422

Sigma-Aldrich

2,2,3,3,4,4,5,5-Octafluoro-1,6-hexanediol

98%

Synonym(s):

1,6-Dihydroxy-2,2,3,3,4,4,5,5-octafluorohexane, 1H ,1H ,6H ,6H -Perfluorohexane-1,6-diol

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$132.00

$132.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
$132.00

About This Item

Linear Formula:
HOCH2(CF2)4CH2OH
CAS Number:
Molecular Weight:
262.10
Beilstein:
1784039
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

$132.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

98%

form

solid

bp

100 °C/3 mmHg (lit.)

mp

66-70 °C (lit.)

SMILES string

OCC(F)(F)C(F)(F)C(F)(F)C(F)(F)CO

InChI

1S/C6H6F8O2/c7-3(8,1-15)5(11,12)6(13,14)4(9,10)2-16/h15-16H,1-2H2

InChI key

NHEKBXPLFJSSBZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service