Skip to Content
Merck
All Photos(1)

Documents

406058

Sigma-Aldrich

2-Fluoro-6-methoxybenzonitrile

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
FC6H3(OCH3)CN
CAS Number:
Molecular Weight:
151.14
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

solid

mp

103-105 °C (lit.)

SMILES string

COc1cccc(F)c1C#N

InChI

1S/C8H6FNO/c1-11-8-4-2-3-7(9)6(8)5-10/h2-4H,1H3

InChI key

YPMSIWYNTPSPMV-UHFFFAOYSA-N

General description

2-Fluoro-6-methoxybenzonitrile (FMBN) is a fluorinated organic building block used in organic synthesis. Molecular structure and vibrational spectra of FMBN have been investigated.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Vibrational spectra and electrostatic potential surface of 2-fluoro-6-methoxybenzonitrile based on quantum chemical calculations.
Murugan M, et al.
Journal of Chemical and Pharmaceutical Research, 4(7), 3400-3413 (2012)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service