Skip to Content
Merck
All Photos(1)

Key Documents

405604

Sigma-Aldrich

3-Nitrofluoranthene

technical grade, 90%

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$97.20

$97.20


Estimated to ship on06 April 2025


Request a Bulk Order

Select a Size

Change View
1 G
$97.20

About This Item

Empirical Formula (Hill Notation):
C16H9NO2
CAS Number:
Molecular Weight:
247.25
Beilstein:
2216474
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$97.20


Estimated to ship on06 April 2025


Request a Bulk Order

grade

technical grade

Quality Level

Assay

90%

form

powder

mp

157-159 °C (lit.)

functional group

nitro

SMILES string

[O-][N+](=O)c1ccc2-c3ccccc3-c4cccc1c24

InChI

1S/C16H9NO2/c18-17(19)15-9-8-13-11-5-2-1-4-10(11)12-6-3-7-14(15)16(12)13/h1-9H

InChI key

PIHGQKMEAMSUNA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

3-Nitrofluoranthene (3-NFA) is a nitro-polycyclic aromatic hydrocarbon (nitro-PAH).[1][2] It is an air pollutant present in diesel exhaust and also in some xerographic toners and copies.[3] Vibrational spectra of 3-NFA has been analyzed by FT-IR and density functional theory calculations.[4] Detection of 3-NFA in suspended particulate matter has been reported by liquid chromatography with fluorescence detection.[2] Carcinogenicity of 3-nitrofluoranthene in male F344/DuCrj rats has been studied.[3] Voltammetric determination of 3-nitrofluoranthene in mixed methanol-water solutions by differential pulse voltammetry (DPV) at boron doped diamond thin-film electrode (BDDE) has been described.[5] Analysis of 3-nitrofluoranthene, by HPLC separation and with electrochemiluminescence detection (ECL) has been reported.[6]

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shan-Hong Lan et al.
Environmental science and pollution research international, 21(12), 7390-7399 (2014-03-01)
Total suspended and size-segregated atmospheric particles were collected in four seasons at three representative points in different functional areas of Dongguan City. The detailed size distributions of six nitro-PAHs [2-nitrofluorene (2-NF), 9-nitroanthracene, 2-nitrofluoranthene (2-NFL), 3-nitrofluoranthene, 1-nitropyrene, and 2-nitropyrene (2-NP)] were
Electrochemiluminescence determination of nitro polycyclic-aromatic hydrocarbons using HPLC separation.
Chen X, et al.
Chromatographia, 58(9-10), 571-577 (2003)
H Ohgaki et al.
Cancer letters, 15(1), 1-7 (1982-01-01)
1-Nitropyrene and 3-nitrofluoranthene are present in diesel exhaust, in pollutants in air, and were also present in certain xerographic toners and copies. Their carcinogenicities were studied in male F344/DuCrj rats by subcutaneous injection. Sarcomas, mainly malignant fibrous histiocytomas at the
Voltammetric Determination of 3-Nitrofluoranthene and 3-Aminofluoranthene at Boron Doped Diamond Thin-Film Electrode.
Cizek K, et al.
Electroanalysis, 19(12), 1295-1299 (2007)
Experimental and theoretical study of vibrational spectra of 3-nitrofluoranthene.
Onchoke KK and Parks M.
Journal of Molecular Structure, 999(1), 22-28 (2011)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service