Skip to Content
Merck
All Photos(1)

Key Documents

40486

Sigma-Aldrich

Heptadecafluoro-1-octanesulfonyl chloride

technical, ≥90% (T)

Synonym(s):

Perfluoro-1-octanesulfonyl chloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8ClF17O2S
CAS Number:
Molecular Weight:
518.58
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

grade

technical

Assay

≥90% (T)

SMILES string

FC(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)S(Cl)(=O)=O

InChI

1S/C8ClF17O2S/c9-29(27,28)8(25,26)6(20,21)4(16,17)2(12,13)1(10,11)3(14,15)5(18,19)7(22,23)24

InChI key

FJHZKRYJOILIGD-UHFFFAOYSA-N

General description

Heptadecafluoro-1-octanesulfonyl chloride is a fluorinated building block.

Application

Heptadecafluoro-1-octanesulfonyl chloride (1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8,8-heptadecafluorooctane-1-sulfonyl chloride) may be used to investigate the substrates having perfluoro leaving groups and their use in labelling chemistry.

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 2 - Carc. 2 - Lact. - Repr. 1B - STOT RE 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Use of perfluoro groups in nucleophilic 18F-fluorination.
Blom E, et al.
Journal of Labelled Compounds & Radiopharmaceuticals, 53(1), 24-30 (2010)
Harshita Krishnatreyya et al.
Expert opinion on drug delivery, 17(6), 863-880 (2020-04-11)
Defense personnel utilize capsaicin-based ocular sprays as non-lethal agents for law implementation during instances of mob violence. This study involves the capsaicin antagonist Capsazepine and the investigation of whether Capsazepine's antagonistic approach can be favorably utilized for defense utilization to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service