Skip to Content
Merck
All Photos(1)

Documents

404675

Sigma-Aldrich

Isophorone oxide

99%

Synonym(s):

2,3-Epoxy-3,5,5-trimethyl-1-cyclohexanone, 2,3-Epoxy-3,5,5-trimethylcyclohexanone, 3,5,5-Trimethyl-2,3-epoxycyclohexanone, 4,4,6-Trimethyl-7-oxabicyclo[4.1.0]heptan-2-one, Epoxyisophorone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H14O2
CAS Number:
Molecular Weight:
154.21
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

Assay

99%

refractive index

n20/D 1.453 (lit.)

bp

75-76 °C/6 mmHg (lit.)

density

0.994 g/mL at 25 °C (lit.)

SMILES string

CC1(C)CC(=O)C2OC2(C)C1

InChI

1S/C9H14O2/c1-8(2)4-6(10)7-9(3,5-8)11-7/h7H,4-5H2,1-3H3

InChI key

ROTNQFPVXVZSRL-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

181.4 °F - closed cup

Flash Point(C)

83 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tetyana Kobets et al.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association, 115, 228-243 (2018-03-20)
Genotoxicity of flavor and fragrance materials was assessed in Turkey Egg Genotoxicity Assay (TEGA) using 32P-nucleotide postlabeling (NPL) and comet assays to detect hepatic DNA adducts and strand breaks. Twenty materials having results in GADD45a-Gluc 'BlueScreen HC' genotoxicity assay, and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service