402702
2,2,2,3′-Tetrafluoroacetophenone
99%
Synonym(s):
α,α,α,3-Tetrafluoroacetophenone
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
FC6H4COCF3
CAS Number:
Molecular Weight:
192.11
Beilstein:
1872788
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Assay
99%
form
liquid
refractive index
n20/D 1.444 (lit.)
bp
79 °C/50 mmHg (lit.)
density
1.372 g/mL at 25 °C (lit.)
SMILES string
Fc1cccc(c1)C(=O)C(F)(F)F
InChI
1S/C8H4F4O/c9-6-3-1-2-5(4-6)7(13)8(10,11)12/h1-4H
InChI key
NQSPTMFCJGKOQJ-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
General description
2,2,2,3′-Tetrafluoroacetophenone (m-fluorotrifluoroacetophenone, α,α,α,3-Tetrafluoroacetophenone) is a fluorine substituted aryl alkyl ketone. Electron transfer quenching of m-fluorotrifluoroacetophenone has been studied.
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
3 - Flammable liquids
WGK
WGK 3
Flash Point(F)
62.6 °F - closed cup
Flash Point(C)
17 °C - closed cup
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Electron transfer quenching of fluorine substituted aryl alkyl ketones. Fluorine-19 polarization via the triplet mechanism.
Manion Schilling ML, et al.
Journal of the American Chemical Society, 99(24), 7792-7799 (1977)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service