Skip to Content
Merck
All Photos(1)

Documents

400793

Sigma-Aldrich

Niobium(V) iodide

−8 mesh

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
NbI5
CAS Number:
Molecular Weight:
727.43
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:

form

solid

particle size

−8 mesh

mp

>300 °C (lit.)

SMILES string

I[Nb](I)(I)(I)I

InChI

1S/5HI.Nb/h5*1H;/q;;;;;+5/p-5

InChI key

FWIYBTVHGYLSAZ-UHFFFAOYSA-I

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service