Skip to Content
Merck
All Photos(1)

Key Documents

40000

Sigma-Aldrich

trans-1,4-Dimethylcyclohexane

≥98.0% (GC)

Synonym(s):

trans-Hexahydro-p-xylene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H16
CAS Number:
Molecular Weight:
112.21
Beilstein:
1900344
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

≥98.0% (GC)

form

liquid

refractive index

n20/D 1.421

bp

117-119 °C

solubility

water: soluble

density

0.761 g/mL at 20 °C (lit.)

SMILES string

C[C@H]1CC[C@H](C)CC1

InChI

1S/C8H16/c1-7-3-5-8(2)6-4-7/h7-8H,3-6H2,1-2H3/t7-,8-

InChI key

QRMPKOFEUHIBNM-ZKCHVHJHSA-N

Looking for similar products? Visit Product Comparison Guide

General description

trans-1,4-Dimethylcyclohexane is a branched cyclohexane (C6) compound. Rate coefficients for the reaction of Cl atoms and OH radicals with trans-1,4-dimethylcyclohexane has been evaluated by Fourier transform infrared (FTIR) spectroscopy and gas chromatography-mass spectrometry (GC-MS).[1] Oxidation of trans-1,4-dimethylcyclohexane with H2O2 catalyzed by hexafluorophosphate salt and carboxylic acid has been reported.[2] Crystal structure of trans-1,4-Dimethylcyclohexane has been studied.[3]

Application

trans-1,4-Dimethylcyclohexane (trans-Hexahydro-p-xylene) has been used to evaluate its solid-liquid phase transitions upto pressures of 300MPa and temperatures of 293K. Experimental data for the transition has been represented by Simon equation and 2nd order polynomial equation.[4]

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

42.8 °F - closed cup

Flash Point(C)

6 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Investigation on the solidification of several pure cyclic and aromatic hydrocarbons at pressures to 300MPa.
Wu Y, et al.
Fuel: The Science and Technology of Fuel and Energy, 111, 75-80 (2013)
Rahana Abd Rahman et al.
International journal of molecular sciences, 21(7) (2020-04-09)
In preeclampsia, widespread maternal endothelial dysfunction is often secondary to excessive generation of placental-derived anti-angiogenic factors, including soluble fms-like tyrosine kinase-1 (sFlt-1) and soluble endoglin (sEng), along with proinflammatory cytokines such as tumour necrosis factor-α (TNF-α) and activin A, understanding
Kim-Hung Huynh et al.
International journal of molecular sciences, 21(11) (2020-06-11)
Histamine intoxication associated with seafood consumption represents a global health problem. The consumption of high concentrations of histamine can cause illnesses ranging from light symptoms, such as a prickling sensation, to death. In this study, gold-silver alloy-embedded silica (SiO2@Au@Ag) nanoparticles
Martin Rothe et al.
Scientific reports, 9(1), 3859-3859 (2019-03-09)
Metal nanoparticles are the most frequently used nanostructures in plasmonics. However, besides nanoparticles, metal nanowires feature several advantages for applications. Their elongation offers a larger interaction volume, their resonances can reach higher quality factors, and their mode structure provides better
Yixue Sun et al.
Veterinary microbiology, 223, 9-20 (2018-09-04)
H9N2 avian influenza viruses are of significance in poultry and public health for the past two decades. Vaccination plays an important role in preventing the infection in domestic poultry. Current H9N2 vaccines have not yet offered ideal protection and eliminated

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service