399779
2,2,2-Trifluoro-N-[(S)-α-methylbenzyl]acetamide
99%
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
CF3CONHCH(CH3)C6H5
CAS Number:
Molecular Weight:
217.19
MDL number:
UNSPSC Code:
12352002
PubChem Substance ID:
Assay
99%
optical activity
[α]20/D −140±5°, c = 1 in chloroform
mp
93-95 °C (lit.)
SMILES string
C[C@H](NC(=O)C(F)(F)F)c1ccccc1
InChI
1S/C10H10F3NO/c1-7(8-5-3-2-4-6-8)14-9(15)10(11,12)13/h2-7H,1H3,(H,14,15)/t7-/m0/s1
InChI key
ZJFCMJDNHPXGBY-ZETCQYMHSA-N
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service