Skip to Content
Merck
All Photos(2)

Key Documents

398799

Sigma-Aldrich

Dichloro(1,10-phenanthroline)palladium(II)

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$212.00

$212.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
$212.00

About This Item

Empirical Formula (Hill Notation):
C12H8Cl2N2Pd
CAS Number:
Molecular Weight:
357.53
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22

$212.00


Check Cart for Availability

Request a Bulk Order

reaction suitability

core: palladium
reaction type: Buchwald-Hartwig Cross Coupling Reaction
reaction type: Cross Couplings
reaction type: Heck Reaction
reaction type: Hiyama Coupling
reaction type: Negishi Coupling
reaction type: Sonogashira Coupling
reaction type: Stille Coupling
reaction type: Suzuki-Miyaura Coupling
reagent type: catalyst

SMILES string

Cl[Pd]Cl.c1cnc2c(c1)ccc3cccnc23

InChI

1S/C12H8N2.2ClH.Pd/c1-3-9-5-6-10-4-2-8-14-12(10)11(9)13-7-1;;;/h1-8H;2*1H;/q;;;+2/p-2

InChI key

YGAISFRMWCCXCG-UHFFFAOYSA-L

Application

Catalyst for:
  • Carbonylation of alkyl nitrites
  • Arylation reactions
  • Multiple acetylene insertions under Sonogashira reaction protocol
  • Cross-coupling reactions of pyridine carboxylic acid chlorides with alkylzinc reagents
  • Hydrophosphinylation of alkenes and alkynes
  • Heck reaction
  • Regioselective and stereoselective aminohalogenation reactions
  • Reaction of chloroenynes and chlorodienes with Grignard reagents for the synthesis of stereodefined enynes and dienes

  • Affects the aggregation of human prion protein PrP106-126

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service