Skip to Content
Merck
All Photos(1)

Documents

398322

Sigma-Aldrich

Chloroplatinic acid hydrate

99.9% trace metals basis

Synonym(s):

Hexachloroplatinic(IV) acid hydrate, Hydrogen hexachloroplatinate(IV) hydrate, Platinic chloride hydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2PtCl6 · xH2O
CAS Number:
Molecular Weight:
409.81 (anhydrous basis)
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:

Assay

99.9% trace metals basis

composition

Pt, 38-40%

mp

60 °C (lit.)

density

2.43 g/mL at 25 °C (lit.)

SMILES string

O.Cl.Cl.Cl[Pt](Cl)(Cl)Cl

InChI

1S/6ClH.H2O.Pt/h6*1H;1H2;/q;;;;;;;+4/p-4

InChI key

SLIOYUPLNYLSSR-UHFFFAOYSA-J

Looking for similar products? Visit Product Comparison Guide

Application

A powerful hydrosilylation catalyst for preparation of trans-β-silanes.

replaced by

Product No.
Description
Pricing

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Eye Dam. 1 - Resp. Sens. 1 - Skin Corr. 1B - Skin Sens. 1

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Denmark et al.
Organic letters, 2(4), 565-568 (2000-05-18)
[reaction: see text] Alkenylsilanols bearing methyl ((E)-1 and (Z)-1) or isopropyl ((E)-2 and (Z)-2)) substituents are converted to disubstituted alkenes by a palladium(0)-catalyzed cross-coupling reaction with aryl or vinyl iodides in the presence of tetrabutylammonium fluoride or hydroxide. Yields and
Chun-Yuan Wang et al.
Nature communications, 6, 7734-7734 (2015-07-16)
The development of ultrasmooth, macroscopic-sized silver (Ag) crystals exhibiting reduced losses is critical to fully characterize the ultimate performance of Ag as a plasmonic material, and to enable cascaded and integrated plasmonic devices. Here we demonstrate the growth of single-crystal
C Haris Saslis-Lagoudakis et al.
Scientific reports, 5, 11942-11942 (2015-07-15)
The global herbal products market has grown in recent years, making regulation of these products paramount for public healthcare. For instance, the common horsetail (Equisetum arvense L.) is used in numerous herbal products, but it can be adulterated with closely

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service