Skip to Content
Merck
All Photos(2)

Key Documents

395382

Sigma-Aldrich

Vanadium(III) chloride tetrahydrofuran complex (1:3)

97%

Synonym(s):

Trichlorotris(tetrahydrofuran)vanadium

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$130.00
25 G
$386.00

$130.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
$130.00
25 G
$386.00

About This Item

Linear Formula:
VCl3 · 3C4H8O
CAS Number:
Molecular Weight:
373.62
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
form:
powder

$130.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

97%

form

powder

reaction suitability

reagent type: catalyst
core: vanadium

mp

265 °C (dec.) (lit.)

application(s)

battery manufacturing

storage temp.

2-8°C

SMILES string

Cl[V](Cl)Cl.C1CCOC1

InChI

1S/C4H8O.3ClH.V/c1-2-4-5-3-1;;;;/h1-4H2;3*1H;/q;;;;+3/p-3

InChI key

WARDLUZTYLPMGJ-UHFFFAOYSA-K

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Flam. Sol. 2 - Skin Corr. 1B

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service