Skip to Content
Merck

394785

Sigma-Aldrich

Calcium hexaboride

−200 mesh, 99.5%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CaB6
CAS Number:
Molecular Weight:
104.94
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:

Assay

99.5%

form

powder

particle size

−200 mesh

density

2.3 g/mL at 25 °C (lit.)

SMILES string

[Ca++].B12B3[B-]14B5B4[B-]235

InChI

1S/B6.Ca/c1-2-5(1)3-4(5)6(1,2)3;/q-2;+2

InChI key

AWVIDOFQNLOVRC-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service