Skip to Content
Merck
All Photos(2)

Documents

394394

Sigma-Aldrich

Samarium(III) oxide

99.999% trace rare earth metals basis

Synonym(s):

Samaria

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Sm2O3
CAS Number:
EC Number:
MDL number:
UNSPSC Code:
12352303
PubChem Substance ID:
NACRES:
NA.23

Assay

99.999% trace rare earth metals basis

form

powder

reaction suitability

reagent type: catalyst
core: samarium

density

8.35 g/mL at 25 °C (lit.)

SMILES string

O=[Sm]O[Sm]=O

InChI

1S/3O.2Sm

InChI key

PRCWVHVINXAFRG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Matthew D Burke et al.
Pharmaceutical research, 24(4), 695-704 (2007-03-21)
To develop a robust radiolabeling technique to enable evaluation of difficult to radiolabel gastric retentive formulations using gamma scintigraphy. The use of a successful radiolabel will allow accurate assessment of the gastric residence time of the formulations. The retention of
Janne Marvola et al.
International journal of pharmaceutics, 281(1-2), 3-10 (2004-08-04)
This paper is a report from a pharmacoscintigraphic study with an Egalet constant-release system containing caffeine and natural abundance samarium oxide. First the formulation was tested in vitro to clarify integrity during irradiation in the nuclear reactor. Then six healthy
B Mandal et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 60(4), 933-939 (2004-03-24)
Microphone based photoacoustic (PA) spectrometer to study solids in powder form was designed and fabricated. Laser PA spectra of Sm3+ ion in Sm2O3 and SmCl3.6H2O microcrystalline powders were recorded first time in 484-542 nm spectral region at room temperature. Analysis
S F Ahrabi et al.
European journal of pharmaceutical sciences : official journal of the European Federation for Pharmaceutical Sciences, 8(3), 193-201 (1999-06-24)
The effect of the neutron activation factors, i.e., admixture of samarium oxide (Sm2O3) and irradiation time, on the physico-chemical properties of the raw materials and the in vitro dissolution and disintegration of hydrophilic and lipophilic suppositories was investigated. It was
Lloyd G Tillman et al.
Journal of pharmaceutical sciences, 97(1), 225-236 (2007-08-28)
Treatment of systemic disease with phosphorothioate antisense oligonucleotides (PS ASOs) has been accomplished using local or parenteral routes of administration to date. This report describes, for the first time, the effective oral delivery of a second generation oligonucleotide where significant

Articles

Rechargeable solid-state batteries are becoming increasingly important due to wide-spread use in computers, portable electronics, and vehicular applications.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service