Skip to Content
Merck
All Photos(1)

Key Documents

393568

Sigma-Aldrich

α-Bromo-α-methyl-γ-butyrolactone

96%

Sign Into View Organizational & Contract Pricing

Select a Size

25 ML
$540.00

$540.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 ML
$540.00

About This Item

Empirical Formula (Hill Notation):
C5H7BrO2
CAS Number:
Molecular Weight:
179.01
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$540.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

96%

form

liquid

refractive index

n20/D 1.494 (lit.)

bp

112 °C/13 mmHg (lit.)

density

1.581 g/mL at 25 °C (lit.)

functional group

bromo
ester

SMILES string

CC1(Br)CCOC1=O

InChI

1S/C5H7BrO2/c1-5(6)2-3-8-4(5)7/h2-3H2,1H3

InChI key

MXEVTHILFFSWOG-UHFFFAOYSA-N

General description

α-Bromo-α-methyl-γ-butyrolactone is a lactone derivative. It has been reported to be hydrolyzed by paraoxonase/arylesterase (PON1), a purified rabbit serum.[1]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

D I Draganov et al.
The Journal of biological chemistry, 275(43), 33435-33442 (2000-08-10)
The paraoxonase gene family contains at least three members: PON1, PON2, and PON3. The physiological roles of the corresponding gene products are still uncertain. Until recently, only the serum paraoxonase/arylesterase (PON1) had been purified and characterized. Here we report the

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service