Skip to Content
Merck
All Photos(1)

Key Documents

393436

Sigma-Aldrich

Tris(2,6-dimethoxyphenyl)phosphine

98%

Synonym(s):

DMPP

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[(CH3O)2C6H3]3P
CAS Number:
Molecular Weight:
442.44
MDL number:
UNSPSC Code:
12352112
PubChem Substance ID:

Assay

98%

form

powder

reaction suitability

reaction type: Buchwald-Hartwig Cross Coupling Reaction
reaction type: Heck Reaction
reaction type: Hiyama Coupling
reaction type: Negishi Coupling
reaction type: Sonogashira Coupling
reaction type: Stille Coupling
reaction type: Suzuki-Miyaura Coupling
reagent type: ligand
reaction type: Cross Couplings

mp

145-147 °C (lit.)

functional group

phosphine

SMILES string

COc1cccc(OC)c1P(c2c(OC)cccc2OC)c3c(OC)cccc3OC

InChI

1S/C24H27O6P/c1-25-16-10-7-11-17(26-2)22(16)31(23-18(27-3)12-8-13-19(23)28-4)24-20(29-5)14-9-15-21(24)30-6/h7-15H,1-6H3

InChI key

CMLWFCUAXGSMBB-UHFFFAOYSA-N

Application

Catalyst for:
  • Preparation of chiral building blocks via hydroalkynylation reactions or ynolates
  • Three-component aza-Morita-Baylis-Hillman reactions (aza-MBH)
  • Atom-economic synthesis of nitrogen heterocycles and ynenoates from alkynes
  • Oxycylizations of allendiols
  • Enantioselective aldol reactions
  • Pd-mediated cross-coupling, Pd- and Cu-mediated benzannulation reactions and copper-mediated dimerization reactions

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Marloes van Hout et al.
Biochemical pharmacology, 174, 113788-113788 (2019-12-31)
α6β2-Containing nicotinic acetylcholine receptors (α6β2* nAChRs) are predominantly expressed in midbrain dopaminergic neurons, including substantia nigra pars compacta (SNc) neurons and their projections to striatal regions, where they regulate dopamine release and nigrostriatal activity. It is well established that nAChR
Evangelia S Papadopoulou et al.
Frontiers in microbiology, 11, 581283-581283 (2020-12-01)
Nitrification inhibitors (NIs) applied to soil reduce nitrogen fertilizer losses from agro-ecosystems. NIs that are currently registered for use in agriculture appear to selectively inhibit ammonia-oxidizing bacteria (AOB), while their impact on other nitrifiers is limited or unknown. Ethoxyquin (EQ)
Jamal Sheikhi et al.
Ecotoxicology and environmental safety, 191, 110243-110243 (2020-02-01)
The use of nitrification inhibitors (NIs) such as 3,4-dimethylpyrazole phosphate (DMPP) has been suggested to diminish agricultural soil nitrate (NO3-) loss and increase nitrogen (N) use efficiency (NUE). However, the yield of ammonium (NH4+)-sensitive plants such as spinach (Spinacia oleracea
José Moya-Díaz et al.
Acta physiologica (Oxford, England), 228(4), e13417-e13417 (2019-11-27)
It is widely accepted that the exocytosis of synaptic and secretory vesicles is triggered by Ca2+ entry through voltage-dependent Ca2+ channels. However, there is evidence of an alternative mode of exocytosis induced by membrane depolarization but lacking Ca2+ current and
Jeningsih et al.
Sensors (Basel, Switzerland), 20(7) (2020-03-29)
A DNA micro-optode for dengue virus detection was developed based on the sandwich hybridization strategy of DNAs on succinimide-functionalized poly(n-butyl acrylate) (poly(nBA-NAS)) microspheres. Gold nanoparticles (AuNPs) with an average diameter of ~20 nm were synthesized using a centrifugation-based method and

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service