Skip to Content
Merck
All Photos(1)

Key Documents

393258

Sigma-Aldrich

Bis(2-carbopentyloxy-3,5,6-trichlorophenyl) oxalate

technical grade

Synonym(s):

CPPO

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[CO2C6H(Cl)3CO2(CH2)4CH3]2
CAS Number:
Molecular Weight:
677.18
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

grade

technical grade

Assay

≥96.5% (by AgNO3, titration)

form

solid

mp

76-80 °C (lit.)

SMILES string

CCCCCOC(=O)c1c(Cl)cc(Cl)c(Cl)c1OC(=O)C(=O)Oc2c(Cl)c(Cl)cc(Cl)c2C(=O)OCCCCC

InChI

1S/C26H24Cl6O8/c1-3-5-7-9-37-23(33)17-13(27)11-15(29)19(31)21(17)39-25(35)26(36)40-22-18(14(28)12-16(30)20(22)32)24(34)38-10-8-6-4-2/h11-12H,3-10H2,1-2H3

InChI key

PURKHUDOTFUVNG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Bis(2-carbopentyloxy-3,5,6-trichlorophenyl) oxalate is a peroxyoxalate based material that is used as in chemiluminescent dye which can further be utilized as a detection method for liquid phase analysis. It can be solubilized in organic solvents to achieve high quantum efficiencies and produce a strong chemiluminescent blue light.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Chemiluminescent properties of copolyesters containing red and blue chromophores.
Lee CW, et al.
Synthetic Metals, 126(1), 97-104 (2002)
Chemiluminescent properties of blue fluorophores containing naphthalene unit.
Cheon JW, et al.
Dyes and Pigments, 61(1), 23-30 (2004)
Improved microfluidic chip-based sequential-injection trapped-droplet array liquid-liquid extraction system for determination of aluminium.
Shen H and Fang Q
Talanta, 77(1), 269-272 (2008)
Thin-film organic photodiodes as integrated detectors for microscale chemiluminescence assays.
Hofmann O, et al.
Sensors and Actuators B, Chemical, 106(2), 878-884 (2005)

Articles

Graphene is the building block for carbon nanomaterials with different dimensionalities.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service