Skip to Content
Merck
All Photos(1)

Documents

391387

Sigma-Aldrich

4-Amino-5-chloro-2,6-dimethylpyrimidine

96%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H8ClN3
CAS Number:
Molecular Weight:
157.60
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

96%

mp

164-166 °C (lit.)

SMILES string

Cc1nc(C)c(Cl)c(N)n1

InChI

1S/C6H8ClN3/c1-3-5(7)6(8)10-4(2)9-3/h1-2H3,(H2,8,9,10)

InChI key

BKFCZKYCVQQMCX-UHFFFAOYSA-N

General description

4-Amino-5-chloro-2,6-dimethylpyrimidine (ACDP) is an aminopyrimidine derivative. Its interaction as an electron donor with a σ-electron acceptor, iodine has been studied.
4-Amino-5-chloro-2,6-dimethylpyrimidine participates in the synthesis of diazino[c]quinolin-5(6H)-one and -isoquinolin-6(5H)-one, diazino[c]naphthyridin-6(5H)- and -5(6H)-one derivatives.

Application

4-Amino-5-chloro-2,6-dimethylpyrimidine is suitable reagent employed as a nucleobase standard in the systematic quantitative structure-retention relationship (QSRR) study on pyrimidines between the molecular descriptors (variables) and their chromatographic retention (experimental unit).

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis of substituted diazino[c] quinolin-5(6H)-ones, diazino[c]isoquinolin-6(5H)-ones, diazino [c]naphthyridin-6(5H)-ones and diazino[c]naphthyridin-5(6H)-ones.
Fresneau N, et al.
Tetrahedron, 69(26), 5393-5400 (2013)
Usama M Rabie et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 68(3), 605-611 (2007-02-24)
Interactions of some pyrimidine derivatives, 4-amino-2,6-dimethylpyrimidine, kyanmethin, (4AP), 2-amino-4,6-dimethylpyrimidine (2AP), 2-aminopyrimidine (AP), 2-amino-4-methylpyrimidine (AMP), 2-amino-4-methoxy-6-methylpyrimidine (AMMP), and 4-amino-5-chloro-2,6-dimethylpyrimidine (ACDP) as electron donors, with iodine (I(2)), as a typical sigma-electron acceptor, have been studied. Electronic absorption spectra of these interactions in
Multivariate Chemometric Study on the Interfacial Properties of Nucleic-Acid Bases.
Luo HB, et al.
Journal of Data Science, 1(4), 447-460 (2003)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service