Skip to Content
Merck
All Photos(1)

Key Documents

389455

Sigma-Aldrich

Bromoethane-d5

99 atom % D

Synonym(s):

Deuterated bromoethane, Ethyl-d5 bromide, Pentadeuteroethyl bromide

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$132.00
5 G
$292.00

$132.00


Check Cart for Availability

Request more information

Select a Size

Change View
1 G
$132.00
5 G
$292.00

About This Item

Linear Formula:
CD3CD2Br
CAS Number:
Molecular Weight:
114.00
Beilstein:
1698455
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.12

$132.00


Check Cart for Availability

Request more information

vapor pressure

7.96 psi ( 20 °C)

Quality Level

isotopic purity

99 atom % D

Assay

99% (CP)

form

liquid

refractive index

n20/D 1.421 (lit.)

bp

37-40 °C (lit.)

mp

-119 °C (lit.)

density

1.527 g/mL at 25 °C

mass shift

M+5

storage temp.

2-8°C

SMILES string

[2H]C([2H])([2H])C([2H])([2H])Br

InChI

1S/C2H5Br/c1-2-3/h2H2,1H3/i1D3,2D2

InChI key

RDHPKYGYEGBMSE-ZBJDZAJPSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Carc. 2 - Flam. Liq. 2 - Ozone 1

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

-9.4 °F

Flash Point(C)

-23 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service