Skip to Content
Merck
All Photos(1)

Key Documents

389366

Sigma-Aldrich

1,3-Dinitrobenzene-d4

98 atom % D

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$88.00

$88.00


Estimated to ship on13 April 2025


Request more information

Select a Size

Change View
1 G
$88.00

About This Item

Linear Formula:
C6D4(NO2)2
CAS Number:
Molecular Weight:
172.13
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.12

$88.00


Estimated to ship on13 April 2025


Request more information

isotopic purity

98 atom % D

Quality Level

form

solid

bp

297 °C (lit.)

mp

88-90 °C (lit.)

mass shift

M+4

SMILES string

[2H]c1c([2H])c(c([2H])c(c1[2H])[N+]([O-])=O)[N+]([O-])=O

InChI

1S/C6H4N2O4/c9-7(10)5-2-1-3-6(4-5)8(11)12/h1-4H/i1D,2D,3D,4D

InChI key

WDCYWAQPCXBPJA-RHQRLBAQSA-N

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Signal Word

Danger

Hazard Classifications

Acute Tox. 1 Dermal - Acute Tox. 2 Inhalation - Acute Tox. 2 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - STOT RE 2

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

302.0 °F - closed cup

Flash Point(C)

150.00 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service