Skip to Content
Merck
All Photos(1)

Key Documents

388076

Sigma-Aldrich

tert-Butyl peracetate solution

50 wt. % in odorless mineral spirits

Synonym(s):

tert -Butyl peracetate, tert -Butyl peroxyacetate

Sign Into View Organizational & Contract Pricing

Select a Size

500 ML
$151.00

$151.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
500 ML
$151.00

About This Item

Linear Formula:
CH3CO3C(CH3)3
CAS Number:
Molecular Weight:
132.16
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

$151.00


Check Cart for Availability

Request a Bulk Order

form

liquid

t1/2

10 hr(101 °C, 0.2 M in benzene)

concentration

50 wt. % in odorless mineral spirits

refractive index

n20/D 1.412

density

0.828 g/mL at 25 °C

SMILES string

CC(=O)OOC(C)(C)C

InChI

1S/C6H12O3/c1-5(7)8-9-6(2,3)4/h1-4H3

InChI key

SWAXTRYEYUTSAP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Allows the direct copper catalyzed acetoxylation of β-lactams at the 4-position.[1]
Reagent used as a methyl-radical source under photoredox catalysis in conjunction with photocatalyst [Ir{dF(CF3)ppy}2(dtbpy)]PF6 (Aldrich L511765) for late-stage functionalization of biologically active heterocycles.

Late-Stage Functionalization of Biologically Active Heterocycles Through Photoredox Catalysis

Product can be used with our line of photoreactors: Including Penn PhD (Z744035) & SynLED 2.0 (Z744080)

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Asp. Tox. 1 - Carc. 1B - Eye Irrit. 2 - Flam. Liq. 3 - Muta. 1B - Org. Perox. C - Skin Irrit. 2 - STOT SE 3

Target Organs

Central nervous system, Respiratory system

Storage Class Code

5.2 - Organic peroxides and self-reacting hazardous materials

WGK

WGK 3

Flash Point(F)

98.6 °F - closed cup

Flash Point(C)

37 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Journal of the Chemical Society. Perkin Transactions 1, 277-277 (1990)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service