Skip to Content
Merck
All Photos(1)

Key Documents

387703

Sigma-Aldrich

Poly(Bisphenol A-co-epichlorohydrin), glycidyl end-capped

average Mn ~355

Synonym(s):

Phenol,4,4′-(1-methylethylidene)bis-,polymerwith2,2′-[(1-methylethylidene)bis(4,1-phenyleneoxymethylene)]bis[oxirane]

Sign Into View Organizational & Contract Pricing

Select a Size

100 ML
$58.50

$58.50


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 ML
$58.50

About This Item

CAS Number:
MDL number:
UNSPSC Code:
12162002
NACRES:
NA.23

$58.50


Check Cart for Availability

Request a Bulk Order

form

liquid

mol wt

average Mn ~355

viscosity

50-65 poise(25 °C)

transition temp

Tg (DSC) −18 °C (onset)

density

1.168 g/mL at 25 °C

SMILES string

ClCC4OC4.O3C(C3)CO.Oc1ccc(cc1)C(C)(C)c2ccc(cc2)O.OC(CO)CO

InChI key

YCDGZAHDCFWTHG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

  • Self assembling healing agents for mendable epoxy networks: This study discusses the application of glycidyl end-capped poly(Bisphenol A-co-epichlorohydrin) as healing agents in epoxy networks, highlighting their self-assembling capabilities for enhanced material recovery (FR Jones et al., 2014).
  • Nanoscience and Nanotechnology Research: This paper covers the integration of glycidyl end-capped poly(Bisphenol A-co-epichlorohydrin) with white Portland cement to create nanocomposites, exploring the effects of these composites on material properties (A Kausar, 2015).
  • Improving the Recyclability of an Epoxy Resin through the Addition of New Biobased Vitrimer: This study focuses on the addition of a biobased vitrimer to glycidyl end-capped poly(bisphenol A-co-epichlorohydrin), analyzing its impact on the thermal and mechanical properties of the epoxy resin (A Veloso-Fernández et al., 2023).

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Sens. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service